Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GCH1
Entrez Gene ID
396146
Gene Name
GTP cyclohydrolase 1
Gene Aliases
-
Chromosome Location
Chr.5: 56530543 - 56547351 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_205223.1 NP_990554.1 2 - 3 450 149
GenBank mRNA Z49267.1 - - 450 149

View Full Product Details