Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDX3X
Entrez Gene ID
418567
Gene Name
DEAD-box helicase 3, X-linked
Gene Aliases
CPL10
Chromosome Location
Chr.1: 113327064 - 113344500 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030800.1 NP_001025971.1 12 - 13 1405 115
XM_015301535.2 XP_015157021.1 12 - 13 1401 115
GenBank mRNA AJ851409.1 - 12 - 13 1405 115
MH119574.1 - 12 - 13 1292 115

View Full Product Details