Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DDX3X
Entrez Gene ID
418567
Gene Name
DEAD-box helicase 3, X-linked
Gene Aliases
CPL10
Chromosome Location
Chr.1: 113327064 - 113344500 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030800.1 NP_001025971.1 4 - 5 397 72
XM_015301535.2 XP_015157021.1 4 - 5 393 72
GenBank mRNA AJ851409.1 - 4 - 5 397 72
MH119574.1 - 4 - 5 284 72

View Full Product Details