Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
FUT7
Entrez Gene ID
427782
Gene Name
fucosyltransferase 7
Gene Aliases
CFTII
Chromosome Location
Chr.17: 815128 - 821035 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_015279614.2 XP_015135100.1 2 - 2 786 77
XM_025141317.1 XP_024997085.1 2 - 2 3656 77
XM_025141318.1 XP_024997086.1 2 - 2 3576 77

View Full Product Details