Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZC3H3
Entrez Gene ID
430349
Gene Name
zinc finger CCCH-type containing 3
Gene Aliases
-
Chromosome Location
Chr.2: 148657720 - 148813448 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031585.2 NP_001026756.1 12 - 13 2875 69
XM_015283175.2 XP_015138661.1 12 - 13 2944 69
GenBank mRNA AJ851559.1 - 12 - 13 2876 69

View Full Product Details