Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H3
Entrez Gene ID
430349
Gene Name
zinc finger CCCH-type containing 3
Gene Aliases
-
Chromosome Location
Chr.2: 148657720 - 148813448 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031585.2 NP_001026756.1 4 - 5 1776 59
XM_015283175.2 XP_015138661.1 4 - 5 1842 59
GenBank mRNA AJ851559.1 - 4 - 5 1777 59

View Full Product Details