Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SEPT5
Entrez Gene ID
416778
Gene Name
septin 5
Gene Aliases
SEP-05, septin-5
Chromosome Location
Chr.15: 1035336 - 1075936 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030654.2 NP_001025825.2 4 - 5 280 102
XM_015275241.2 XP_015130727.1 3 - 4 468 102
XM_025155436.1 XP_025011204.1 4 - 5 4039 102
GenBank mRNA AJ720671.1 - 4 - 5 285 102
AJ851383.1 - 4 - 5 280 102

View Full Product Details