The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001005843.1 | NP_001005843.1 | 1 - 2 | 274 | 63 |
GenBank mRNA | AJ719669.1 | - | 1 - 2 | 274 | 63 |