Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SDF4
Entrez Gene ID
419423
Gene Name
stromal cell derived factor 4
Gene Aliases
-
Chromosome Location
Chr.21: 2535907 - 2548707 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001006302.1 NP_001006302.1 4 - 5 721 153
GenBank mRNA AJ720139.1 - 4 - 5 721 153

View Full Product Details