Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
HOXA6
Entrez Gene ID
420630
Gene Name
homeobox A6
Gene Aliases
HOX1, HOX1.2, HOX1B
Chromosome Location
Chr.2: 32626024 - 32628685 on Build GRCg6a
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001030987.2 NP_001026158.1 1 - 1 342 69
GenBank mRNA AY307380.1 - 1 - 1 348 69

View Full Product Details