Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
WRKY56
Entrez Gene ID
100170678
Gene Name
transcription factor
Gene Aliases
-
Chromosome Location
Chr.8: 17848938 - 17850671 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001250629.1 NP_001237558.1 2 - 3 233 75
GenBank mRNA AK245181.1 - 2 - 3 252 75
BT094603.1 - 2 - 3 253 75
EU375348.1 - 2 - 3 233 75

View Full Product Details