Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ACS1
Entrez Gene ID
547976
Gene Name
1-aminocyclopropane 1-carboxylate synthase
Gene Aliases
-
Chromosome Location
Chr.5: 41020530 - 41023024 on Build Glycine_max_v2.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001249929.1 NP_001236858.1 4 - 4 1641 99
GenBank mRNA AK244524.1 - 2 - 2 1084 99
X67100.1 - 4 - 4 1641 99

View Full Product Details