Gene Transcripts

Gene Symbol
CDR2
Entrez Gene ID
1039
Gene Name
cerebellar degeneration related protein 2
Gene Aliases
CDR62, Yo
Chromosome Location
Chr. 16: 22345936 - 22374617 on Build GRCh38
UniGene ID
Hs.513430
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001802.1 NP_001793.1 5 963
GenBank mRNA AK292273.1 - 5 710
BC017503.1 - 5 945 4873337
D12981.1 - 5 639
M63256.1 - 5 823

View Full Product Details