Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001146218.1 | NP_001139690.1 | 4 | 382 | |
NM_004627.4 | NP_004618.2 | 4 | 492 | ||
GenBank mRNA | AK293113.1 | - | 4 | 447 | |
AK293866.1 | - | 4 | 425 | ||
AK299144.1 | - | 4 | 335 | ||
AK310268.1 | - | 4 | 996 | ||
BC012415.1 | - | 4 | 425 | 3865141 | |
BC111393.1 | - | 4 | 381 | ||
DB498697.1 | - | 4 | 383 | ||
Y12478.1 | - | 4 | 436 |