Gene Transcripts

Gene Symbol
CCNB2
Entrez Gene ID
9133
Gene Name
cyclin B2
Gene Aliases
HsT17299
Chromosome Location
Chr. 15: 59105085 - 59125045 on Build GRCh38
UniGene ID
Hs.194698
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004701.3 NP_004692.1 2 233
GenBank mRNA AB020981.1 - 2 118
AF002822.1 - 2 68
AF087910.1 - 2 175
AK001404.1 - 2 197
AK222611.1 - 2 166
AL080146.1 - 2 186
AM851054.1 - 2 49
BC105086.1 - 2 181 8144089
BC105112.1 - 2 181 8144115
BT019458.1 - 2 49
BU934242.1 - 2 234
CR533527.1 - 2 49

View Full Product Details