Gene Transcripts

Gene Symbol
OR12D2
Entrez Gene ID
26529
Gene Name
olfactory receptor family 12 subfamily D member 2 (gene/pseudogene)
Gene Aliases
DJ994E9.8, HS6M1-20
Chromosome Location
Chr. 6: 29396639 - 29397671 on Build GRCh38
UniGene ID
Hs.247862
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013936.3 NP_039224.2 1 499
GenBank mRNA BC069123.1 - 1 438
BC101742.1 - 1 499 8069248
BC101746.1 - 1 499 8069252

View Full Product Details