Gene Transcripts

Gene Symbol
NUDT9
Entrez Gene ID
53343
Gene Name
nudix hydrolase 9
Gene Aliases
NUDT10
Chromosome Location
Chr. 4: 87420002 - 87459454 on Build GRCh38
UniGene ID
Hs.149500
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001248011.1 NP_001234940.1 5, 6 1017
NM_024047.4 NP_076952.1 6, 7 1113
NM_198038.2 NP_932155.1 6, 7 755
XM_011532032.2 XP_011530334.1 6, 7 3292
GenBank mRNA AF265341.1 - 6, 7 754
AF273028.1 - 6, 7 1077
AK027295.1 - 6, 7 1103
AK074845.1 - 6, 7 755
AK075408.1 - 6, 7 1033
AK308028.1 - 5, 6 784
AY026252.1 - 6, 7 1108
AY359123.1 - 6, 7 807
BC000542.2 - 6, 7 873 3163944
BQ446599.1 - 2, 3 607

View Full Product Details