Gene Transcripts

Gene Symbol
CMC2
Entrez Gene ID
56942
Gene Name
C-X9-C motif containing 2
Gene Aliases
2310061C15Rik, C16orf61, DC13
Chromosome Location
Chr. 16: 80975800 - 81007046 on Build GRCh38
UniGene ID
Hs.55028
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020188.3 NP_064573.1 2 250
XM_017023461.1 XP_016878950.1 2 381
XM_017023462.1 XP_016878951.1 2 396
XM_017023463.1 XP_016878952.1 2 378
XM_017023464.1 XP_016878953.1 2 379
XM_017023465.1 XP_016878954.1 2 388
XM_017023467.1 XP_016878956.1 4 333
XM_017023468.1 XP_016878957.1 3 243
XM_017023469.1 XP_016878958.1 3 317
XM_017023470.1 XP_016878959.1 3 241
GenBank mRNA AF201935.1 - 2 226
AK123193.1 - 1 1605
BC032631.1 - 2 254 5399193

View Full Product Details