Gene Transcripts

Gene Symbol
HIVEP3
Entrez Gene ID
59269
Gene Name
human immunodeficiency virus type I enhancer binding protein 3
Gene Aliases
KBP-1, KBP1, KRC, SHN3, Schnurri-3, ZAS3, ZNF40C
Chromosome Location
Chr. 1: 41506365 - 42035925 on Build GRCh38
UniGene ID
Hs.403972
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001127714.2 NP_001121186.1 3 5213
NM_024503.4 NP_078779.2 4 5293
XM_011541884.2 XP_011540186.1 5 5225
XM_011541885.2 XP_011540187.1 4 5505
XM_011541886.2 XP_011540188.1 4 5505
XM_011541887.2 XP_011540189.1 4 5505
XM_017001992.1 XP_016857481.1 5 6472
XM_017001993.1 XP_016857482.1 3 5425
XM_017001994.1 XP_016857483.1 4 5505
GenBank mRNA AB046775.3 - 3 5213
AF278765.1 - 4 5086
BC152563.1 - 3 5213 8860523

View Full Product Details