Gene Transcripts

Gene Symbol
CTBP2
Entrez Gene ID
1488
Gene Name
C-terminal binding protein 2
Gene Aliases
Chromosome Location
Chr. 10: 124987849 - 125161061 on Build GRCh38
UniGene ID
Hs.501345
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083914.2 NP_001077383.1 6 869
NM_001290214.2 NP_001277143.1 6 854
NM_001290215.2 NP_001277144.1 6 1118
NM_001321012.1 NP_001307941.1 6 813
NM_001321013.1 NP_001307942.1 5 1014
NM_001321014.1 NP_001307943.1 6 1115
NM_001329.3 NP_001320.1 6 857
NM_022802.2 NP_073713.2 4 2174
XM_005269561.2 XP_005269618.1 7 1040
XM_005269564.2 XP_005269621.1 7 1248
XM_005269567.2 XP_005269624.1 7 1247
XM_005269568.4 XP_005269625.1 7 975
XM_005269569.2 XP_005269626.1 6 936
XM_005269571.2 XP_005269628.1 5 695
XM_005269572.3 XP_005269629.1 5 743
XM_006717642.2 XP_006717705.1 8 1169
XM_011539349.2 XP_011537651.1 4 951
XM_011539351.1 XP_011537653.1 9 1270
XM_011539353.1 XP_011537655.1 8 1141
XM_011539354.1 XP_011537656.1 7 1065
XM_011539355.1 XP_011537657.1 6 1143
XM_011539357.1 XP_011537659.1 6 881
XM_011539358.1 XP_011537660.1 5 959
XM_017015756.1 XP_016871245.1 7 911
XM_017015757.1 XP_016871246.1 6 2637
GenBank mRNA AF016507.1 - 6 774
AF222711.1 - 4 2191
AK290390.1 - 6 718
AL833398.1 - 6 829
BC002486.2 - 6 830
BC037900.2 - 4 838 5276086
BC047018.1 - 6 834
BC052276.1 - 6 811
BC072020.1 - 6 791
BM800106.1 - 4 529
BT007012.1 - 4 429
CV029885.1 - 4 429
CX165167.1 - 6 683
DB200827.1 - 4 402

View Full Product Details