Gene Transcripts

Gene Symbol
C8orf82
Entrez Gene ID
414919
Gene Name
chromosome 8 open reading frame 82
Gene Aliases
Chromosome Location
Chr. 8: 144526219 - 144529074 on Build GRCh38
UniGene ID
Hs.105685
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001795.1 NP_001001795.1 3 689
GenBank mRNA BC064373.1 - 3 689
BC064829.1 - 3 621 6176032
BC073936.1 - 2 943 5223300

View Full Product Details