Gene Transcripts

Gene Symbol
NRARP
Entrez Gene ID
441478
Gene Name
NOTCH-regulated ankyrin repeat protein
Gene Aliases
Chromosome Location
Chr. 9: 137299631 - 137302251 on Build GRCh38
UniGene ID
Hs.535075
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004354.2 NP_001004354.1 1 449
GenBank mRNA BC053618.1 - 1 449

View Full Product Details