Gene Transcripts
- Gene Symbol
- TARM1
- Entrez Gene ID
- 441864
- Gene Name
- T cell-interacting, activating receptor on myeloid cells 1
- Gene Aliases
- OLT-2
- Chromosome Location
- Chr. 19: 54069342 - 54081365 on Build GRCh38
- UniGene ID
- Hs.694674
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK301730.1 |
- |
3 |
634 |
|
DQ479398.1 |
- |
3 |
349 |
|
View Full Product Details