Gene Transcripts

Gene Symbol
ARMC3
Entrez Gene ID
219681
Gene Name
armadillo repeat containing 3
Gene Aliases
CT81, KU-CT-1
Chromosome Location
Chr. 10: 22928024 - 23038527 on Build GRCh38
UniGene ID
Hs.659807
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001282745.1 NP_001269674.1 6 510
NM_001282746.1 NP_001269675.1 6 510
NM_173081.4 NP_775104.2 6 510
XM_005252380.2 XP_005252437.1 6 493
XM_005252381.2 XP_005252438.1 5 331
XM_011519350.2 XP_011517652.1 6 493
XM_017015831.1 XP_016871320.1 6 493
XM_017015832.1 XP_016871321.1 6 493
XM_017015833.1 XP_016871322.1 6 493
GenBank mRNA AK057389.1 - 6 453
AK090961.1 - 4 454
AK302985.1 - 6 497
AY436643.1 - 6 471
BC039312.1 - 6 503 5296302

View Full Product Details