Gene Transcripts

Gene Symbol
TRAM1
Entrez Gene ID
23471
Gene Name
translocation associated membrane protein 1
Gene Aliases
PNAS8, TRAM, TRAMP
Chromosome Location
Chr. 8: 70573218 - 70608459 on Build GRCh38
UniGene ID
Hs.491988
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001317804.1 NP_001304733.1 10 964
NM_001317805.1 NP_001304734.1 9 1479
NM_014294.5 NP_055109.1 9 1047
XM_017013266.1 XP_016868755.1 8 919
XM_017013267.1 XP_016868756.1 8 1263
GenBank mRNA AK289397.1 - 9 907
AK292777.1 - 9 831
AK303411.1 - 10 892
BC000687.2 - 9 869 3347823
BC020283.1 - 9 862
BC037738.1 - 9 959 5261490
BT007359.1 - 9 787
CR541654.1 - 9 787
CR541739.1 - 9 787
X63679.1 - 9 908

View Full Product Details