Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001014436.2 | NP_001014436.1 | 3 | 272 | |
NM_001122956.1 | NP_001116428.1 | 3 | 272 | ||
NM_014063.6 | NP_054782.2 | 3 | 272 | ||
XM_011515325.1 | XP_011513627.1 | 3 | 243 | ||
GenBank mRNA | AF077353.1 | - | 3 | 212 | |
AF151364.1 | - | 3 | 184 | ||
AF197060.1 | - | 3 | 174 | ||
AF218020.3 | - | 3 | 206 | ||
AF250287.1 | - | 3 | 174 | ||
AK027367.1 | - | 3 | 201 | ||
AK293151.1 | - | 3 | 210 | ||
AK293279.1 | - | 3 | 239 | ||
AK302037.1 | - | 3 | 256 | ||
BC011677.2 | - | 3 | 195 | 4299867 | |
BC031687.1 | - | 3 | 180 | 5168099 | |
CR457167.1 | - | 3 | 174 | ||
DC404852.1 | - | 3 | 272 |