Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001014436.2 | NP_001014436.1 | 10 | 1003 | |
NM_001122956.1 | NP_001116428.1 | 10 | 1030 | ||
NM_001284313.1 | NP_001271242.1 | 8 | 834 | ||
NM_001284315.1 | NP_001271244.1 | 8 | 818 | ||
NM_014063.6 | NP_054782.2 | 10 | 1006 | ||
XM_011515325.1 | XP_011513627.1 | 10 | 998 | ||
GenBank mRNA | AB209486.1 | - | 8 | 690 | |
AF077353.1 | - | 10 | 943 | ||
AF151364.1 | - | 10 | 915 | ||
AF197060.1 | - | 10 | 905 | ||
AF218020.3 | - | 10 | 937 | ||
AF250287.1 | - | 10 | 905 | ||
AK027367.1 | - | 10 | 959 | ||
AK126142.1 | - | 6 | 983 | ||
AK293151.1 | - | 9 | 869 | ||
AK293279.1 | - | 9 | 826 | ||
AK293340.1 | - | 9 | 822 | ||
AK293698.1 | - | 8 | 799 | ||
AK296778.1 | - | 9 | 755 | ||
AK298102.1 | - | 9 | 779 | ||
AK300630.1 | - | 8 | 738 | ||
AK302037.1 | - | 10 | 990 | ||
BC011677.2 | - | 10 | 929 | 4299867 | |
BC031687.1 | - | 10 | 911 | 5168099 | |
CR457167.1 | - | 10 | 905 |