Gene Transcripts

Gene Symbol
OR51D1
Entrez Gene ID
390038
Gene Name
olfactory receptor family 51 subfamily D member 1
Gene Aliases
OR11-14, OR51D1Q
Chromosome Location
Chr. 11: 4639715 - 4640838 on Build GRCh38
UniGene ID
Hs.553728
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001004751.2 NP_001004751.1 1 756
GenBank mRNA BC140717.1 - 1 756

View Full Product Details