Gene Transcripts

Gene Symbol
MS4A12
Entrez Gene ID
54860
Gene Name
membrane spanning 4-domains A12
Gene Aliases
Ms4a10
Chromosome Location
Chr. 11: 60492743 - 60507430 on Build GRCh38
UniGene ID
Hs.272789
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001164470.1 NP_001157942.1 5, 6 618
NM_017716.2 NP_060186.2 6, 7 756
XM_011545117.2 XP_011543419.1 7, 8 904
GenBank mRNA AK000224.1 - 6, 7 756

View Full Product Details