Gene Transcripts

Gene Symbol
CDC37L1
Entrez Gene ID
55664
Gene Name
cell division cycle 37 like 1
Gene Aliases
CDC37B, HARC
Chromosome Location
Chr. 9: 4679553 - 4708399 on Build GRCh38
UniGene ID
Hs.666439
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017913.3 NP_060383.2 5 880
GenBank mRNA AK000497.1 - 5 875
AK000646.1 - 5 764
AK225510.1 - 5 764
BC014133.2 - 5 700 4622495

View Full Product Details