Gene Transcripts
- Gene Symbol
- FCRL2
- Entrez Gene ID
- 79368
- Gene Name
- Fc receptor like 2
- Gene Aliases
- CD307b, FCRH2, IFGP4, IRTA4, SPAP1, SPAP1A, SPAP1B, SPAP1C
- Chromosome Location
- Chr. 1: 157745733 - 157777472 on Build GRCh38
- UniGene ID
- Hs.437393
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_030764.3 |
NP_110391.2
|
11, 12 |
1555 |
|
NR_125358.1 |
|
7, 8 |
637 |
|
XM_011509976.2 |
XP_011508278.1
|
10, 11 |
1825 |
|
GenBank mRNA |
AF319438.1 |
- |
6, 7 |
1040 |
|
AF390037.1 |
- |
6, 7 |
878 |
|
AF459633.1 |
- |
11, 12 |
1543 |
|
AL833194.1 |
- |
6, 7 |
3414 |
|
AY043465.1 |
- |
11, 12 |
1542 |
|
AY358130.1 |
- |
11, 12 |
1545 |
|
BC069185.1 |
- |
7, 8 |
641 |
|
BC129836.1 |
- |
6, 7 |
880 |
40115446 |
View Full Product Details