Gene Transcripts

Gene Symbol
NDNF
Entrez Gene ID
79625
Gene Name
neuron derived neurotrophic factor
Gene Aliases
C4orf31, NORD
Chromosome Location
Chr. 4: 121035627 - 121072518 on Build GRCh38
UniGene ID
Hs.709520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_024574.3 NP_078850.3 4 2191
GenBank mRNA AK026844.1 - 1 1278
AY358821.1 - 4 1763
BC019351.1 - 4 2087 3928768

View Full Product Details