Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_004738.4 | NP_004729.1 | 3 | 609 | |
GenBank mRNA | AF086628.1 | - | 3 | 415 | |
AF160212.1 | - | 3 | 307 | ||
AK023241.1 | - | 3 | 606 | ||
AK123538.1 | - | 3 | 612 | ||
AY358464.1 | - | 3 | 501 | ||
BC001712.1 | - | 4 | 446 | 3543354 | |
BT009811.1 | - | 3 | 268 |