Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 88416377-88416911 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA7
Entrez Gene ID
55812
Gene Name
spermatogenesis associated 7
Gene Aliases
HEL-S-296, HSD-3.1, HSD3, LCA3
UniGene
Hs.525518
Gene Chromosome Location
Chr.14: 88385638-88470350 on Build GRCh38
Transcripts
NM_001040428.3 NM_018418.4 XM_005267851.1 XM_005267852.1 XM_005267854.1 XM_005267855.1 XM_006720204.1 XM_006720205.1 XM_011536951.1 XM_011536952.1 XM_011536953.1 XM_017021452.1 XM_017021453.1 XM_017021454.1 XM_017021455.1 XM_017021456.1 XM_017021457.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7115908 c.357T>A p.F119L Chr.14: 88416829-88416829 on Build GRCh38
1749011 c.325G>A p.A109T Chr.14: 88416797-88416797 on Build GRCh38
2250296 c.274G>A p.E92K Chr.14: 88416746-88416746 on Build GRCh38
2250299 c.364T>A p.L122I Chr.14: 88416836-88416836 on Build GRCh38
7411815 c.350C>T p.S117L Chr.14: 88416822-88416822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370110205 Chr.14: 88416795-88416795 on Build GRCh38
rs747880531 Chr.14: 88416664-88416664 on Build GRCh38
rs1018690355 Chr.14: 88416659-88416659 on Build GRCh38
rs762001916 Chr.14: 88416875-88416875 on Build GRCh38
rs752911477 Chr.14: 88416710-88416710 on Build GRCh38
rs972509901 Chr.14: 88416606-88416606 on Build GRCh38
rs201229698 Chr.14: 88416409-88416409 on Build GRCh38
rs767275913 Chr.14: 88416715-88416715 on Build GRCh38
rs868213711 Chr.14: 88416654-88416654 on Build GRCh38
rs1005866862 Chr.14: 88416556-88416556 on Build GRCh38
rs777598239 Chr.14: 88416755-88416755 on Build GRCh38
rs754923626 Chr.14: 88416817-88416817 on Build GRCh38
rs781466391 Chr.14: 88416859-88416859 on Build GRCh38
rs775594191 Chr.14: 88416788-88416788 on Build GRCh38
rs777346333 Chr.14: 88416736-88416739 on Build GRCh38
rs373779457 Chr.14: 88416815-88416815 on Build GRCh38
rs763053429 Chr.14: 88416791-88416791 on Build GRCh38
rs1048878519 Chr.14: 88416445-88416445 on Build GRCh38
rs61747004 Chr.14: 88416756-88416756 on Build GRCh38
rs539976712 Chr.14: 88416518-88416518 on Build GRCh38
rs556939845 Chr.14: 88416673-88416673 on Build GRCh38
rs761053219 Chr.14: 88416678-88416678 on Build GRCh38
rs1000075430 Chr.14: 88416555-88416555 on Build GRCh38
rs1016789232 Chr.14: 88416567-88416567 on Build GRCh38
rs769850431 Chr.14: 88416786-88416786 on Build GRCh38
rs199825301 Chr.14: 88416726-88416726 on Build GRCh38
rs80044281 Chr.14: 88416794-88416794 on Build GRCh38
rs746642029 Chr.14: 88416851-88416851 on Build GRCh38
rs747784419 Chr.14: 88416737-88416737 on Build GRCh38
rs35137272 Chr.14: 88416829-88416829 on Build GRCh38
rs749310044 Chr.14: 88416868-88416868 on Build GRCh38
rs917372270 Chr.14: 88416759-88416759 on Build GRCh38
rs1022789201 Chr.14: 88416528-88416528 on Build GRCh38
rs202230167 Chr.14: 88416836-88416836 on Build GRCh38
rs1332533 Chr.14: 88416433-88416433 on Build GRCh38
rs3903242 Chr.14: 88416488-88416488 on Build GRCh38
rs891941122 Chr.14: 88416408-88416408 on Build GRCh38
rs773863529 Chr.14: 88416874-88416874 on Build GRCh38
rs147016354 Chr.14: 88416409-88416409 on Build GRCh38
rs759599766 Chr.14: 88416697-88416697 on Build GRCh38
rs370952510 Chr.14: 88416784-88416784 on Build GRCh38
rs569509558 Chr.14: 88416567-88416570 on Build GRCh38
rs772554840 Chr.14: 88416669-88416669 on Build GRCh38
rs767745816 Chr.14: 88416760-88416760 on Build GRCh38
rs140287375 Chr.14: 88416725-88416725 on Build GRCh38
rs1045786317 Chr.14: 88416493-88416493 on Build GRCh38
rs200034171 Chr.14: 88416782-88416782 on Build GRCh38
rs770278373 Chr.14: 88416788-88416788 on Build GRCh38
rs142011483 Chr.14: 88416542-88416542 on Build GRCh38
rs766157792 Chr.14: 88416797-88416797 on Build GRCh38
rs950129104 Chr.14: 88416781-88416781 on Build GRCh38
rs550144579 Chr.14: 88416873-88416873 on Build GRCh38
rs777506416 Chr.14: 88416845-88416845 on Build GRCh38
rs746361238 Chr.14: 88416861-88416861 on Build GRCh38
rs182231428 Chr.14: 88416521-88416521 on Build GRCh38
rs910123180 Chr.14: 88416724-88416724 on Build GRCh38
rs1054661201 Chr.14: 88416400-88416400 on Build GRCh38
rs374511557 Chr.14: 88416870-88416870 on Build GRCh38
rs1037148646 Chr.14: 88416512-88416512 on Build GRCh38
rs763218291 Chr.14: 88416712-88416712 on Build GRCh38
rs895033855 Chr.14: 88416546-88416546 on Build GRCh38
rs984361190 Chr.14: 88416693-88416693 on Build GRCh38
rs907220752 Chr.14: 88416496-88416496 on Build GRCh38
rs755765600 Chr.14: 88416697-88416697 on Build GRCh38
rs376502749 Chr.14: 88416717-88416717 on Build GRCh38
rs757636745 Chr.14: 88416855-88416855 on Build GRCh38
rs530016766 Chr.14: 88416863-88416863 on Build GRCh38
rs187297010 Chr.14: 88416661-88416661 on Build GRCh38
rs748824159 Chr.14: 88416741-88416745 on Build GRCh38
rs1028923377 Chr.14: 88416883-88416883 on Build GRCh38
rs145863932 Chr.14: 88416568-88416568 on Build GRCh38
rs778645976 Chr.14: 88416663-88416663 on Build GRCh38
rs575092657 Chr.14: 88416635-88416635 on Build GRCh38
rs985098324 Chr.14: 88416689-88416689 on Build GRCh38
rs748783358 Chr.14: 88416660-88416660 on Build GRCh38
rs766511222 Chr.14: 88416852-88416852 on Build GRCh38
rs778627627 Chr.14: 88416732-88416732 on Build GRCh38
rs73323512 Chr.14: 88416451-88416451 on Build GRCh38
rs771425592 Chr.14: 88416679-88416679 on Build GRCh38
rs150093878 Chr.14: 88416838-88416838 on Build GRCh38
rs754516908 Chr.14: 88416730-88416730 on Build GRCh38
rs536916579 Chr.14: 88416410-88416410 on Build GRCh38
rs765236407 Chr.14: 88416706-88416706 on Build GRCh38
rs955045170 Chr.14: 88416688-88416688 on Build GRCh38

View Full Product Details