The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001104.3 | NP_001095.2 | 11 - 12 | 1375 | 79 |
NM_001258371.2 | NP_001245300.2 | 11 - 12 | 1523 | 79 | |
NR_047663.2 | 11 - 12 | 1375 | 79 | ||
GenBank mRNA | AK125851.1 | - | 11 - 12 | 1909 | 79 |
AK303044.1 | - | 11 - 12 | 1523 | 79 | |
AK313556.1 | - | 11 - 12 | 1313 | 79 | |
BC096323.2 | - | 5 - 6 | 435 | 79 | |
BC099647.3 | - | 11 - 12 | 1296 | 79 | |
BC099649.3 | - | 11 - 12 | 1296 | 79 | |
M86407.1 | - | 11 - 12 | 1296 | 79 |