Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BHMT
Entrez Gene ID
635
Gene Name
betaine--homocysteine S-methyltransferase
Gene Aliases
BHMT1, HEL-S-61p
Chromosome Location
Chr.5: 79111781 - 79132290 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001713.2 NP_001704.2 2 - 3 273 116
GenBank mRNA AK315224.1 - 2 - 3 245 116
BC012616.1 - 2 - 3 249 116
DA370221.1 - 2 - 3 274 116
EU794592.1 - 2 - 3 273 116
U50929.1 - 2 - 3 194 116

View Full Product Details