The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256414.1 | NP_001243343.1 | 3 - 4 | 503 | 123 |
NM_002069.5 | NP_002060.4 | 3 - 4 | 640 | 123 | |
GenBank mRNA | AF055013.1 | - | 3 - 4 | 315 | 123 |
AF493905.1 | - | 3 - 4 | 303 | 123 | |
AK292953.1 | - | 3 - 4 | 614 | 123 | |
AK304442.1 | - | 3 - 4 | 503 | 123 | |
AL049933.1 | - | 3 - 4 | 618 | 123 | |
BC026326.1 | - | 3 - 4 | 639 | 123 | |
BT019775.1 | - | 3 - 4 | 303 | 123 | |
M17219.1 | - | 3 - 4 | 288 | 123 |