The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_000948.5 | NP_000939.1 | 4 - 5 | 1013 | 76 |
NM_001163558.2 | NP_001157030.1 | 5 - 6 | 685 | 76 | |
XM_011514753.2 | XP_011513055.1 | 5 - 6 | 732 | 76 | |
XM_011514754.2 | XP_011513056.1 | 6 - 7 | 745 | 76 | |
GenBank mRNA | BC015850.1 | - | 4 - 5 | 1016 | 76 |
BC088370.1 | - | 4 - 5 | 543 | 76 | |
D00411.1 | - | 3 - 4 | 466 | 76 | |
DQ839492.1 | - | 5 - 6 | 401 | 76 | |
GQ305133.1 | - | 5 - 6 | 685 | 76 | |
M29386.1 | - | 3 - 4 | 466 | 76 | |
U75583.1 | - | 3 - 4 | 465 | 76 | |
V00566.1 | - | 4 - 5 | 498 | 76 | |
X54393.1 | - | 5 - 6 | 635 | 76 |