The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006517.4 | NP_006508.2 | 4 - 5 | 1320 | 146 |
GenBank mRNA | AB085789.1 | - | 4 - 5 | 1262 | 146 |
U05315.1 | - | 4 - 5 | 1352 | 146 |