Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CTNS
Entrez Gene ID
1497
Gene Name
cystinosin, lysosomal cystine transporter
Gene Aliases
CTNS-LSB, PQLC4
Chromosome Location
Chr.17: 3636391 - 3663103 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001031681.2 NP_001026851.2 3 - 4 530 70
NM_004937.2 NP_004928.2 3 - 4 653 70
XM_005256485.2 XP_005256542.1 3 - 4 653 70
XM_006721463.2 XP_006721526.1 3 - 4 602 70
XM_006721464.2 XP_006721527.1 3 - 4 653 70
XM_011523691.2 XP_011521993.1 3 - 4 452 70
XM_017024255.1 XP_016879744.1 3 - 4 653 70
GenBank mRNA AJ222967.1 - 3 - 4 399 70
AK292019.1 - 3 - 4 366 70
AK309651.1 - 3 - 4 363 70
BC032850.2 - 3 - 4 519 70
CR542058.1 - 1 - 2 60 70
KU178003.1 - 1 - 2 60 70
KU178004.1 - 1 - 2 60 70
KU178005.1 - 1 - 2 60 70
KU178006.1 - 1 - 2 60 70

View Full Product Details