Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PSMD5
Entrez Gene ID
5711
Gene Name
proteasome 26S subunit, non-ATPase 5
Gene Aliases
S5B
Chromosome Location
Chr.9: 120798318 - 120843021 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270427.1 NP_001257356.1 4 - 5 659 63
NM_005047.3 NP_005038.1 5 - 6 788 63
XM_011518866.2 XP_011517168.1 5 - 6 698 63
XM_017014927.1 XP_016870416.1 5 - 6 698 63
GenBank mRNA AK026925.1 - 1 - 2 91 63
AK291051.1 - 5 - 6 689 63
AK303007.1 - 4 - 5 589 63
BC014478.1 - 5 - 6 677 63
D31889.1 - 5 - 6 675 63
S79862.1 - 5 - 6 679 63

View Full Product Details