The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001032999.2 | NP_001028171.1 | 3 - 4 | 565 | 70 |
NM_001039709.1 | NP_001034798.1 | 3 - 4 | 695 | 70 | |
NM_005093.3 | NP_005084.1 | 4 - 5 | 983 | 70 | |
XM_011529101.2 | XP_011527403.1 | 3 - 4 | 614 | 70 | |
XM_011529102.2 | XP_011527404.1 | 3 - 4 | 432 | 70 | |
XM_011529103.2 | XP_011527405.1 | 3 - 4 | 514 | 70 | |
XM_017028121.1 | XP_016883610.1 | 3 - 4 | 432 | 70 | |
XM_017028122.1 | XP_016883611.1 | 3 - 4 | 619 | 70 | |
XM_017028123.1 | XP_016883612.1 | 3 - 4 | 432 | 70 | |
XM_017028124.1 | XP_016883613.1 | 2 - 3 | 283 | 70 | |
GenBank mRNA | AF013970.1 | - | 3 - 4 | 456 | 70 |
AF039200.1 | - | 2 - 3 | 359 | 70 | |
AF052210.1 | - | 2 - 3 | 385 | 70 | |
AF068266.1 | - | 2 - 3 | 385 | 70 | |
AF069747.1 | - | 5 - 6 | 1044 | 70 | |
AK307887.1 | - | 3 - 4 | 600 | 70 | |
AK314159.1 | - | 4 - 5 | 782 | 70 | |
BC015066.1 | - | 3 - 4 | 729 | 70 | |
BC016298.2 | - | 1 - 2 | 84 | 70 | |
BC040344.2 | - | 3 - 4 | 526 | 70 |