The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318381.1 | NP_001305310.1 | 2 - 3 | 363 | 87 |
NM_001318382.1 | NP_001305311.1 | 2 - 3 | 363 | 87 | |
NM_014425.4 | NP_055240.2 | 2 - 3 | 363 | 87 | |
NR_134606.1 | 2 - 3 | 363 | 87 | ||
GenBank mRNA | AF039217.2 | - | 2 - 3 | 314 | 87 |
AF084367.1 | - | 1 - 2 | 133 | 87 | |
AK304851.1 | - | 2 - 3 | 288 | 87 | |
AK312795.1 | - | 2 - 3 | 305 | 87 | |
BC006370.2 | - | 2 - 3 | 260 | 87 | |
BC041665.1 | - | 2 - 3 | 319 | 87 | |
BC056897.1 | - | 2 - 3 | 282 | 87 | |
BC111761.1 | - | 1 - 2 | 146 | 87 | |
BC112351.1 | - | 1 - 2 | 146 | 87 | |
BP280580.1 | - | 2 - 3 | 328 | 87 | |
DB217652.1 | - | 2 - 3 | 363 | 87 |