Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PIGT
Entrez Gene ID
51604
Gene Name
phosphatidylinositol glycan anchor biosynthesis class T
Gene Aliases
CGI-06, MCAHS3, NDAP, PNH2
Chromosome Location
Chr.20: 45416067 - 45426245 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184729.2 NP_001171658.1 2 - 3 451 68
NM_015937.5 NP_057021.2 2 - 3 451 68
NR_047693.1 2 - 3 451 68
GenBank mRNA AB057724.1 - 2 - 3 361 68
AF132940.1 - 2 - 3 355 68
AK075469.1 - 2 - 3 375 68
AK225517.1 - 2 - 3 381 68
AK293294.1 - 2 - 3 375 68
AL121742.1 - 2 - 3 354 68
AY358588.1 - 2 - 3 358 68
BC015022.1 - 2 - 3 380 68
BC110892.1 - 2 - 3 359 68
BC136827.1 - 2 - 3 389 68
BC136828.1 - 2 - 3 389 68
DA037291.1 - 2 - 3 434 68
DC384944.1 - 2 - 3 441 68
DC399420.1 - 2 - 3 451 68

View Full Product Details