The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001037175.2 | NP_001032252.1 | 3 - 4 | 544 | 76 |
NM_017982.3 | NP_060452.3 | 3 - 4 | 544 | 76 | |
XM_005273169.1 | XP_005273226.1 | 4 - 5 | 561 | 76 | |
XM_006711408.1 | XP_006711471.1 | 3 - 4 | 542 | 76 | |
XM_011509685.1 | XP_011507987.1 | 3 - 4 | 576 | 76 | |
XM_017001583.1 | XP_016857072.1 | 4 - 5 | 742 | 76 | |
XM_017001584.1 | XP_016857073.1 | 4 - 5 | 741 | 76 | |
XM_017001585.1 | XP_016857074.1 | 5 - 6 | 725 | 76 | |
XM_017001586.1 | XP_016857075.1 | 4 - 5 | 619 | 76 | |
XM_017001587.1 | XP_016857076.1 | 4 - 5 | 561 | 76 | |
XM_017001588.1 | XP_016857077.1 | 3 - 4 | 576 | 76 | |
XM_017001589.1 | XP_016857078.1 | 4 - 5 | 746 | 76 | |
GenBank mRNA | AK000914.1 | - | 3 - 4 | 425 | 76 |
AK096265.1 | - | 2 - 3 | 998 | 76 | |
AY358495.1 | - | 3 - 4 | 521 | 76 | |
BC004888.2 | - | 3 - 4 | 425 | 76 | |
EF560748.1 | - | 3 - 4 | 472 | 76 |