The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020639.2 | NP_065690.2 | 5 - 6 | 879 | 92 |
GenBank mRNA | AB047783.1 | - | 5 - 6 | 855 | 92 |
AJ278016.1 | - | 5 - 6 | 879 | 92 | |
AK027424.1 | - | 5 - 6 | 896 | 92 | |
AK296685.1 | - | 3 - 4 | 224 | 92 | |
AK302592.1 | - | 5 - 6 | 690 | 92 | |
AK316412.1 | - | 5 - 6 | 842 | 92 | |
BC035755.1 | - | 5 - 6 | 833 | 92 | |
BC110617.1 | - | 5 - 6 | 852 | 92 | |
BC110618.1 | - | 5 - 6 | 852 | 92 |