Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FIGNL1
Entrez Gene ID
63979
Gene Name
fidgetin like 1
Gene Aliases
-
Chromosome Location
Chr.7: 50437327 - 50450390 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK023142.1 - 2 - 3 202 73

View Full Product Details