The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024074.2 | NP_076979.1 | 1 - 2 | 218 | 58 |
GenBank mRNA | AK025981.1 | - | 1 - 2 | 218 | 58 |
AK292764.1 | - | 1 - 2 | 203 | 58 | |
BC001195.1 | - | 1 - 2 | 134 | 58 |