The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024551.2 | NP_078827.2 | 6 - 7 | 1034 | 60 |
XM_005253789.1 | XP_005253846.1 | 6 - 7 | 1156 | 60 | |
XM_006719018.2 | XP_006719081.1 | 7 - 8 | 1153 | 60 | |
XM_011521024.2 | XP_011519326.1 | 7 - 8 | 1164 | 60 | |
GenBank mRNA | AK025085.1 | - | 3 - 4 | 562 | 60 |
AK127196.1 | - | 7 - 8 | 1089 | 60 | |
AK128511.1 | - | 6 - 7 | 1088 | 60 | |
AY424280.1 | - | 5 - 6 | 913 | 60 | |
BC051858.2 | - | 6 - 7 | 1034 | 60 |