The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001282143.1 | NP_001269072.1 | 6 - 7 | 1908 | 58 |
NM_001282144.1 | NP_001269073.1 | 6 - 7 | 2316 | 58 | |
NM_001282358.1 | NP_001269287.1 | 6 - 7 | 1840 | 58 | |
NM_024618.3 | NP_078894.2 | 6 - 7 | 1935 | 58 | |
XM_005271669.1 | XP_005271726.1 | 6 - 7 | 2159 | 58 | |
XM_005271670.1 | XP_005271727.1 | 5 - 6 | 1812 | 58 | |
XM_005271671.1 | XP_005271728.1 | 5 - 6 | 1986 | 58 | |
XM_005271672.1 | XP_005271729.1 | 5 - 6 | 1809 | 58 | |
XM_006718904.1 | XP_006718967.1 | 6 - 7 | 2193 | 58 | |
XM_011542980.1 | XP_011541282.1 | 6 - 7 | 1823 | 58 | |
GenBank mRNA | AB094095.1 | - | 6 - 7 | 1919 | 58 |
AK025131.1 | - | 1 - 2 | 562 | 58 | |
AK056454.1 | - | 5 - 6 | 1633 | 58 | |
AK095247.1 | - | 4 - 5 | 1120 | 58 | |
AK225223.1 | - | 1 - 2 | 562 | 58 | |
AK291716.1 | - | 6 - 7 | 1902 | 58 | |
AK303002.1 | - | 5 - 6 | 1809 | 58 | |
AY245437.1 | - | 5 - 6 | 1678 | 58 | |
BC013199.1 | - | 2 - 3 | 1039 | 58 | |
BC110890.1 | - | 6 - 7 | 2281 | 58 | |
BX647705.1 | - | 6 - 7 | 1893 | 58 | |
EF452237.1 | - | 5 - 6 | 1678 | 58 |